Solvent Saving Columns
The New HALO® 1.5 - Defining a New Dimension in Chromatography
The adoption of UHPLC instrumentation pushed LC separations to a new level of speed and resolution. In the years since, chromatographers continue to push separation limits in response to increasing challenges. Now in 2022 AMT breaks new ground again with a 1.5 mm internal diameter HALO® column to push the boundaries of adopted UHPLC systems. Founded on all of the benefits of Fused-Core® particles, the HALO® 1.5 delivers increased sensitivity and reduced solvent consumption, allowing scientists to experience the benefits of capillary columns without the pains of specialized microflow systems.
Delivering More Performance
- More Sensitivity from conventional UHPLC Systems
- Higher Ionization Efficiencies from LCMS systems
- 50% solvent reduction compared to 2.1 mm ID columns
- Easy to Implement microflow solution
Solvent Saving Columns Literature
HALO® 1.5 MM ID columns:
Small Molecule (90 Å) |
Peptide (160 Å) |
Protein (1000 Å) |
|
C18, 2.7 um | ES-C18, 2.7 um | C4, 2.7 um | |
AQ-C18, 2.7 um | ES-C18, 2 um | Diphenyl, 2.7 um | |
PCS C18, 2.7 um | PCS C18, 2.7 um | ||
LPH C18, 2.7 um | |||
Penta-HILIC, 2.7 um |