Analytical Sample Prep
Critical decisions are often based on the results of your analytical QC testing. Confidence in those results comes from using products that are proven to perform as expected.
With high-quality filters for sample preparation and dissolution testing, Pall is able to meet the unique requirements of every lab with a wide variety of membranes including the new wwPTFE membrane “the next generation in universal membrane filtration” as well as Nylon, PVDF, hydrophobic PTFE membranes, and more.
The Next Generation Syringe Filter
The new Acrodisc One™ is the next generation syringe filter for aqueous and aggressive organic solvent-based solutions. Extend column life up to 52 times with up to 24% less filter extractables than the competition.
Simplify Mobile Phase Filtration with SolVac®
Simplify Clean-up and Degassing of Mobile Phase Solvents. With a versatile, durable and leak-proof design, draw directly from HPLC Solvent bottles for less spill risk, breakage, and minimal risk of contamination.
Acrodisc MS certified Syringe Filters
The Acrodisc MS is the first syringe filter that is LCMS-certified. The filters are designed for and certified to be very low in LCMS extractables providing more accurate results to the end user.
Ion Chromatography (IC) Acrodisc Syringe Filters
Pall’s Acrodisc Ion Chromatography (IC) syringe filters have been tested to monitor for inorganic extractables. For ion chromatograph applications, only the Acrodisc IC (PES) syringe filter is certified for low levels of inorganic extractables.
Nanosep Centrifugal Devices for Sample Prep
Simple to use and ideal for rapid sample preparation of small volumes, Pall’s Nanosep with new wwPTFE membrane are available in both 0.2 um and 0.45um. Universally compatible with extremely low extractables and binding for the most reliable data.
AcroPrep™ Advance Filter Plates
Pall’s next generation 96-well filter plates have been optimized to support the stringent requirements of high throughput applications, while providing superior performance in a multitude of sample prep applications.