DNA Amplification
NEB offers a wide range of DNA polymerases, and through our commitment to research, ensures the development of innovative, hiqh quality tools for DNA amplification. As the first company to sell Taq DNA Polymerase and the first to discover a PCR-stable high-fidelity DNA polymerase, NEB has established a foundation in developing reliable tools in this space early on. Some more recently developed products include:
- Q5® High-Fidelity DNA Polymerase
- Luna® qPCR and RT-qPCR Products
- OneTaq® DNA Polymerase
- WarmStart® RTx Reverse Transcriptase
Product selection
Q5® High-Fidelity DNA Polymerase
Experience a new standard for fidelity and performance with Q5 High-Fidelity DNA polymerase. Standalone, master mix, and hot start formats available.
Luna® qPCR and RT-qPCR Products
Luna products offer rapid, sensitive, and precise dye- or probe-based qPCR detection and quantitation of target DNA and cDNA sequences.
OneTaq® DNA Polymerase
OneTaq DNA Polymerase offers exceptional performance in endpoint PCR across a wide range of templates. Robust yields with minimal optimization.
WarmStart® RTx Reverse Transcriptase
Ideal for use in isothermal amplification methodologies such as LAMP, WarmStart® RTx enables room temperature setup for increased sensitivity.