BRAND Transferpette Pipettes & Pipette Tips
The newly redesigned BRAND Transferpette S air displacement pipettes have updates to ensure easier, more efficient work. Features a slim grip for comfortable fit in any hand, true one-handed operation, slim shaft for pipetting into narrow tubes, easy cleaning, low operating forces, and universal tip cones to fit tips from most quality manufacturers. Unmatched precision when paired with BRAND pipette tips.
Product selection
NEW Transferpette S Single Channel Pipettes
High quality, air displacement pipettes in volumes from 0.1µL to 10mL.
Transferpette S Multichannel Pipettes
8-channel and 12-channel models in volumes up to 300µL.
BRAND Standard Pipette Tips
Manufactured in one of the world's largest cleanroom facilities to ensure consistently high quality.