Analytik Jena UVP Chem/Gel Studio Imagers
Analytik Jena offers a wide range of high-performance BioImaging Systems designed to fit researchers' specifications and budgets. All systems come in two different sizes and are equipped for chemiluminescence and fluorescence applications with a UV transilluminator, RGB LEDs, and white light. Systems are upgradeable for NIR applications with NIR lasers.
Product selection
UVP ChemStudio touch
High-performance gel and blot imager.
UVP ChemStudio PLUS touch
Imager perfect for larger gels and blots with a 25 x 26 cm imaging area.
UVP GelStudio touch
Daily use high-performance gel documentation imager.
UVP GelStudio PLUS touch
High-performance imager for large gels.