Your One-Stop Shop for Sterilization Prep Materials
Rely on us for everything related to pharmaceutical sterilization packaging and innovative, pre-sterilized disposable products for use in aseptic production to keep your operation in compliance.
Autoclave Bags
Choose from Tyvek styles in 1422A, 1421B, and 1073B. More economical SMS, BHD and Medical grade paper versions are also available,
GMP Equipment Covers
Can be used as an autoclavable cover or as an effective means to keep cleaned equipment protected from external contamination sources.
Sterilization Wrap
Choose from Tyvek styles in 1422A, 1421B, and 1073B. More economical versions are also available in SMS and BHD materials.
Pre-Sterilized Disposable Products
A wide range of validated sterile (10-6 SAL) products including cleanroom markers and pens, trash bags, mop bucket liners and self-sealing sterility maintenance bags.
Sterile Cleanroom Facemasks
Innovative designs to prevent facial exposure and goggles from fogging.