NEBNext® Multiplex Oligos for Illumina® (Index Primers Set 1)
Supplier: New England Biolabs (NEB)
NEBNext multiplex oligos for Illumina (index primers set 1) NEBNext multiplex oligos provide adaptors and primers to enable high yield multiplex Illumina library production.
- High efficiency adaptor ligation
- Minimized adaptor-dimer formation
- Includes index primers for library multiplexing
- Increased sample identification specificity
- Includes USER Enzyme
The unique hairpin loop structure of the NEBNext adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. Additional NEBNext multiplex oligos products are available for different indexing strategies.