CentriVap® Cold Traps, –50 °C, 115 V
Supplier: Labconco
8601000 8601100
78007-958EA 6165 USD
78007-958 78007-974
CentriVap® Cold Traps, –50 °C, 115 V
Vacuum Traps
CentriVap Cold Traps are critical components designed to collect evaporated liquid from CentriVap Vacuum Concentrators. By condensing and trapping volatile solvents, they protect the vacuum pump and the laboratory environment from the harmful effects of vapors, fumes, and corrosives, while also ensuring that solvents can be easily collected for analysis or disposal.
  • For samples containing water
These traps are available either as integral components within configured CentriVap concentration systems or separately, making them suitable for a wide range of laboratory applications such as freeze drying and desiccation. Their role in maintaining system efficiency, safety, and solvent recovery makes them an essential part of many lab workflows.