NEBNext® Enzymatic Methyl-seq v2 Kit
Supplier: New England Biolabs (NEB)
E8015L E8015S
77938-732EA 3021.7 USD
77938-732 77938-734
NEBNext® Enzymatic Methyl-seq v2 Kit
Nucleic Acid Reagents Next Generation Sequencing Reagents
NEBNext Enzymatic Methyl-seq (EM-seq) is a high-performance enzyme-based alternative to bisulfite conversion for the identification of 5 mC and 5 hmC.
  • Superior sensitivity of detection of 5mC and 5hmC
  • 0.1 to 200 ng input range
  • Detection of more CpGs with fewer sequencing reads
  • Even GC coverage
  • Index primers supplied separately
Unlike bisulfite conversion, this highly efficient method minimizes DNA damage, resulting in superior detection of methylated cytosines, with fewer sequencing reads. The new NEBNext Enzymatic Methyl-seq v2 Kit has a wider input range (as low as 100 pg) and a faster, more streamlined workflow than the original EM-seq kit.
The NEBNext Enzymatic Methyl-seq v2 Kit includes conversion reagents, library prep reagents and the EM-seq Adaptor. Multiple sets of the required index primers (NEBNext LV Unique Dual Index Primers) are available separately, enabling greater flexibility in multiplexing.