SteriTrue™ United Scientific Sterile Sampling Scoops, Polystyrene
Supplier: United Scientific Supplies
UNST100 UNST101 UNST102 UNST200 UNST201 UNST202
470359-304PK 125 USD
470359-304 470359-306 470359-308 470359-310 470359-312 470359-314
SteriTrue™ United Scientific Sterile Sampling Scoops, Polystyrene
Scoops
Scoops are ideal for the sampling of powder, granules, and pastes.
  • Ergonomically designed with voice-of-customer input
  • Flat bottom surface for stability during weighing
  • Gamma sterilized, individually packaged
  • Lot stamped for traceability
  • PS material meets FDA guidelines and is safe for food, pharma, and other uses
SteriTrue™ sterile sampling scoops are uniquely developed with end-user input. The polystyrene scoops are ergonomically designed for long periods of use, they can hold stated volumes, and each scoop has a flat bottom surface for stability during weighing.
The most unique feature of the scoops is the long handle; the cross ribs add structural support to help prevent breakage, and the bottom curved design fits comfortably in the palm of the hand.