Zipr-Weld™ Evidence Seals
Supplier: THE LYNN PEAVEY COMPANY
66671 66695 66697
470358-974PK 18.2 USD
470358-974 470358-976 470358-978
Zipr-Weld™ Evidence Seals
Educational Materials Forensics Educational Materials Forensics Supplies
Secure evidence from tampering.
  • Seal any evidence container
  • Tamper-evident design
Zipr-Weld™ evidence strips have zippered edges combined with the welding properties of the adhesive that make removal of the seal virtually impossible, while visibly showing signs of un-authorized entry. Use to secure evidence bags, boxes, cans, jars, bottles, even the doors and windows of a crime scene. The strips measure 1⅜×6⅝".