Latent Lifters
Supplier: THE LYNN PEAVEY COMPANY
57404 57406 05770
470358-968PK 53.2 USD
470358-968 470358-970 470358-916
Latent Lifters
Educational Materials Forensics Educational Materials Forensics Supplies
Record full palm or foot prints.
  • Large size for lifting larger impressions
  • Individual sheets, not on a roll
These lifters can be used for collecting latent finger and palm prints, footprint residue, and trace evidence found at crime scenes. Simply peel off the inside liner and place adhesive down. After lifting, affix backing sheet to evidence lifted.