Arson Evidence Cans
Supplier: THE LYNN PEAVEY COMPANY
05813 05814 05815 05971 05972 05973 30114 51212
470358-918PK 29 USD
470358-918 470358-920 470358-922 470358-924 470358-926 470358-928 470358-942 470358-952
Arson Evidence Cans
Educational Materials Forensics Educational Materials Forensics Supplies
Cans and lids for storing arson evidence.
  • Three sizes and two lining options
  • Adhesive labels also available
These clean, all-metal arson cans with air-tight lids are designed to package items possibly contaminated with accelerants. Any accelerants remaining will evaporate within the air-tight compartment and then a laboratory can draw a head space sample for evaluation. Lined cans have a gray epoxy lining. These metal cans are also being used to store cell phones found or left behind at crime scenes. This can be a huge advantage when investigators and/or police are wanting to examine the information (phone numbers, voicemails, etc.) stored in the phone. The thick metal of the can acts like a barrier, blocking any signals or attempts to try and 'reach' the phone, so information that is in the phone cannot be retrieved, deleted, or altered.