Whirl-Pak® Standard Sterilized Sampling Bags
Supplier: NASCO SAMPLING LLC
B00736 B01009 B01020
11216-057CS 1264.52 USD
11216-057 11216-374 11216-410
Whirl-Pak® Standard Sterilized Sampling Bags
Bags Sample Bags
Whirl-Pak® Standard sterilized sampling bags provide a secure, contaminant-free, flexible container that ensures dependable analysis results.
  • Continuous perimeter seal ensures dependable, leak-proof performance for the safe collection, transport, and storage of solid, semi-solid, and liquid samples
  • Post-manufacturing +1 Sterilization process provides reliable, contaminant-free containers with downloadable sterility documentation for compliance and record-keeping
  • High-quality polyethylene blend delivers excellent durability and visibility for accurate sample inspection and handling
  • Perpendicular Tyvek® pull tabs enable hands-free, aseptic sample collection for enhanced safety and integrity
  • Free from R-Nase, D-Nase, Pyrogen, and BPH
  • Flexible temperature performance from –320 to 180 °F (–196 to 82 °C)
Count on Whirl-Pak® products as a proven, economical, and efficient way to collect, store, and transport samples. First to the closure bag market, the Whirl-Pak® Standard sterilized bag is popular in all industry sectors. With built-in, secure closure tabs, these standard bags are ideal for the shipping and storage of all forms of sample material.
Certifications:
Bags meet USDA, FDA, HACCP, and EPA requirements.
Packaging:
Bags are packaged 500 per box.