Powder and Liquid Samplers, IBB
Supplier: GLOBEPHARMA
M1.BB.2.10.SS M1.BB.3.1.SS M1.BB.3.6.SS M1.BB.BD.SD M1.BB.0.1.SD M1.BB.0.2.SD M1.BB.0.3.SD M1.BB.1.1.SS M1.BB.1.5.SS M1.BB.2.1.SS
77887-590EA 2350.89 USD
77887-590 77887-972 77887-350 77888-090 77888-170 77887-002 77888-422 77887-162 77886-996 77887-086
Powder and Liquid Samplers, IBB
Samplers Solid Samplers
Unit-dose and bulk powder, liquid, and semi-solid samplers available in various lengths and configurations.
  • Precision sampling: designed for accurate and repeatable sampling of powders from bulk containers, ensuring representative and consistent samples for quality control and testing
  • Pharmaceutical-grade construction: made from high-quality 316 L stainless steel or FDA-compliant materials, ensuring compatibility with GMP environments and resistance to corrosion or contamination
  • Ease of cleaning and sterilization: engineered for quick disassembly and thorough cleaning, supporting clean-in-place (CIP) and autoclave sterilization procedures
  • Customizable and scalable: can be tailored to specific sampling applications with adjustable volumes and configurations, ideal for R and D, pilot, and production-scale environments
  • Compliance ready: supports regulatory compliance with FDA, USP, and ISO standards, making them suitable for validated sampling protocols in pharmaceutical and cleanroom operations
Global Pharma specializes in high-quality sampling solutions for the pharmaceutical, biotech, and related industries. The product portfolio includes unit dose and bulk powder, liquid, and semi-solid samplers, designed to meet stringent regulatory requirements. These samplers are available in a variety of configurations to ensure precise and contamination-free sample collection.
With a focus on innovation and compliance, Global Pharma’s samplers cater to diverse manufacturing environments, providing single-use and reusable options for enhanced process efficiency and sterility assurance. Their products are widely used in quality control, R and D, and production settings, making them a trusted choice for pharmaceutical manufacturers worldwide.