Karl Fischer Titrators, EVA V1 and V3
Supplier: Mettler Toledo
30869280 30869282
77775-302EA 11233.55 USD
77775-302 77775-304
Karl Fischer Titrators, EVA V1 and V3
Titrators
The EVA V1 and V3 volumetric Karl Fischer (KF) titrators are ideal for efficiently determining the water content of simple liquids, soluble solids, and more challenging insoluble samples.
  • High reliability and efficiency via the FFA™ control algorithm, a fast-response digital sensor, and state-of-the-art dispensing technology
  • 120000 step dispensing resolution
  • Intuitive OneClick™ user interface simplifies operations and provides customizable shortcuts
  • Automated solvent exchange minimizes exposure to hazardous chemicals
  • Seamless LabX™ integration enhances data management and compliance with regulatory requirements
  • Modular design to meet versatile laboratory needs
The EVA KF Titrators are designed for efficient and accurate titration across various applications, including chemistry, food and beverage, and pharmaceuticals. Measure water content in a wide range of samples, with a limit of quantification as low as 0.2 mg of water per sample. Featuring the new FFA™ control algorithm and biamperometric indication, the EVA KF titrators deliver fast and accurate water content results for any application.
The detachable user-friendly touchscreen with OneClick™ interface streamlines workflows and reduces user errors, while robust data mangement capabilities allow for easy storage and analysis of results. Automatic sample detection and titrant dosing minimizes chemical exposure, ensuring a safer, cleaner laboratory.
With a compact design that fits well in any laboratory, the EVA KF titrators combine innovative technology and practicality, making them an essential for reliable titration.
Results speed: Fast Forecasting Amperometric (FFA™) control algorithm
Dispensing resolution: 120000 steps
Limit of Quantification (LOQ): 0.2 mg water per sample (RSD = 1%)
Accessories information:
Boost lab capabilities with advanced accessories and experience wide-ranging analytical flexibility for diverse applications.
Elevate your analysis with our specialized accessories, including thermostable vessels, homogenizer adapters, and drying ovens. These tools simplify the handling of challenging samples, ensuring precise results. Customize your Karl Fischer process with options like varied burette sizes and extended terminal cables for greater convenience and flexibility.
Ordering information:
Homogenizers (used with the V3) are not offered or supplied by Mettler Toledo and an additional adapter cable is required.
Delivery information:
The EVA V1 and V3 come with all components required for titrations including sensor, 5 ml burette, burette drive, large glass titration vessel and solvent manager.