Sorenson™ MultiRack LTS Compatible Filtered Pipette Tips
Supplier: Corning
87940 87950 87960
77775-474CS 425.15 USD
77775-474 77775-476 77775-478
Sorenson™ MultiRack LTS Compatible Filtered Pipette Tips
Pipette Tips
MultiRack LTS compatible filter tips are fully compatible with LTS pipettors and are available sterile in volumes from 20 to 1000 µl.
  • Compatible with LTS pipettors
  • Extended length options available in 20 and 1000 µl
  • Certified RNase/DNase free, human DNA free, and non-pyrogenic
  • Filters feature 20 µm pore size
Certifications:
ISO 13485, ISO 14001.