ig® RNase Inhibitor, Murine, Glycerol-free, Intact Genomics, Inc.
Supplier: INTACT GENOMICS, INC MS
3712GF 3714GF
104090-576EA 190.38 USD
104090-576 104090-578
ig® RNase Inhibitor, Murine, Glycerol-free, Intact Genomics, Inc.
Enzymes
RNase inhibitors are commonly used to inhibit and control RNase contaminants in enzymatic manipulations of RNA. This version is perfect for freeze-drying into diagnostic kits, ensuring stability and performance for point-of-care tests like molecular diagnostics.
  • Optimal for Lyophilization
  • Better compatibility with sensitive applications
  • Integrates easily into various buffers and systems
RNase Inhibitor is a 49.8 KDa recombinant protein of murine origin which specifically inhibits RNases A, B and C by binding noncovalently in a 1:1 ratio with high affinity. Moreover, it does not inhibit polymerase activity. This Inhibitor from Murine is resistance to oxidation compared to the human/porcine inhibitors as it lacks the pair of cysteines found in the human/porcine versions. In addition, it is stable at less than 1 mM DTT concentration.
Certifications:
Manufactured under ISO 13485:2016 Quality Management certified standards.