IG® T4 UvsX DNA Recombinase, Glycerol-free
Supplier: INTACT GENOMICS, INC MS
3562GF 3565GF 3567GF
104090-550EA 375 USD
104090-550 104090-552 104090-556
IG® T4 UvsX DNA Recombinase, Glycerol-free
Enzymes
Homologous recombination is important for the error-free repair of DNA double-strand breaks and for replication fork restart. Recombinases of the RecA/RAD51 family perform the central catalytic role in this process. UvsX recombinase is the RecA/Rad51 ortholog of bacteriophage T4. T4 UvsX DNA Recombinase and other recombinases form presynaptic filaments on ssDNA that are activated to search for homology in dsDNA and to perform DNA strand exchange (1 to 3).
  • High purity
  • Free from detecable nuclease activity
  • Batch-tested in RPA assay
  • Shipping temperature: Dry ice
Optimal for lyophilization: Perfect for freeze-drying into diagnostic kits, ensuring stability and performance for point-of-care tests like molecular diagnostics.
Certifications:
Manufactured under ISO 13485:2016 Quality Management certified standards.