IG® Bsu DNA Polymerase, Glycerol-free
Supplier: INTACT GENOMICS, INC MS
3585GF 3587GF
104090-566EA 340.38 USD
104090-566 104090-570
IG® Bsu DNA Polymerase, Glycerol-free
Enzymes
Bsu DNA Polymerase I, Large Fragment is a product of the Bacillus subtilis DNA polymerase I which lacks the N-terminal exonuclease domain (1 to 296 amino acids). It retains the 5'→ 3' polymerase activity of DNA polymerase I but lacks the 5'→ 3' exonuclease activity. This large fragment also lacks 3'→ 5' exonuclease activity.
  • High purity
  • Tested in RPA assay
  • Lacks 3'→ 5' exonuclease activity
  • Shipping temperature: Dry ice
Optimal for lyophilization: Perfect for freeze-drying into diagnostic kits, ensuring stability and performance for point-of-care tests like molecular diagnostics.
Certifications:
Manufactured under ISO 13485:2016 Quality Management certified standards.