SS320 Phage Display electrocompetent cells
Supplier: INTACT GENOMICS, INC MS
1173-12 1173-24
104090-462PK 248.08 USD
104090-462 104090-464
SS320 Phage Display electrocompetent cells
Competent Cells
Suitable for the production of phage display libraries.
  • >5×1010cfu/µg Electroporation efficiency
  • Non-amber suppressor strain (MC1061F\’)
  • high-voltage electroporation apparatus 16 kV/cm required
Intact Genomics SS320 (MC1061F\’) phage display electrocompetent cells are suitable for protein expression, general cloning, blue/white screening, M13 phage work and phage display protein expression.
Certifications:
Manufactured under ISO 13485:2016 certified quality management system