ig® Thymidine Auxotrophic EHA105ᵀʰʸ Agrobacterium Electrocompetent Cells
Supplier: INTACT GENOMICS, INC MS
1404-10 1404-30
104090-496PK 421.15 USD
104090-496 104090-498
ig® Thymidine Auxotrophic EHA105ᵀʰʸ Agrobacterium Electrocompetent Cells
Competent Cells
Enables development of more efficient plant transformation systems.
  • Requires supplemental thymidine for growth
  • Enables development of more efficient plant transformation systems
  • Reduced bacterial overgrowth during co-cultivation
  • Decreased need for antibiotics
Intact Genomics (ig®) Thymidine Auxotrophic EHA105ᵀʰʸ Agrobacterium Chemically Competent Cells include modifications so that they will not grow unless 50 mg/L of thymidine is added to Minimal medium.
This prevents the bacteria from overgrowing plant tissues when used for plant transformation.
Thymidine-Auxotrophic EHA105ᵀʰʸ Agrobacterium Chemically Competent Cells are optimized for the highest transformation efficiencies. The EHA105ᵀʰʸ strain is useful for transgenic operations of rice, tobacco and other plants. EAH105ᵀʰʸ– thyA knockout mutant derived from EHA105, itself a derivative of A281 (A136/pTiBo542).
Certifications:
Manufactured under ISO 13485:2016 certified quality management system