ig® Methionine Auxotrophic EHA105ᴹᵉᵗ Electrocompetent Agrobacterium
Supplier: INTACT GENOMICS, INC MS
1278-10 1278-30
104090-470PK 271.15 USD
104090-470 104090-472
ig® Methionine Auxotrophic EHA105ᴹᵉᵗ Electrocompetent Agrobacterium
Competent Cells
Enables development of more efficient plant transformation systems.
  • Optimized for the highest transformation efficiencies
  • Requires supplemental methionine for growth
  • Useful for transgenic operations of plants
  • Amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA)
Intact Genomics (ig®) Methionine-Auxotrophic EHA105ᴹᵉᵗ Agrobacterium Electrocompetent cells include modifications so that they will not grow unless free methionine is added to Minimal medium.
This prevents the bacteria from overgrowing plant tissues when used for plant transformation.
Methionine- Auxotrophic EHA105ᴹᵉᵗ Agrobacterium ElectroCompetent Cells are optimized for the highest transformation efficiencies. The EHA105ᴹᵉᵗ strain is useful for transgenic operations of rice, tobacco and other plants. EHA105 contains a rifampicin resistance gene (rif). EHA105ᴹᵉᵗ strain also contains an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) without self-transport function, which contains the vir gene.
Certifications:
Manufactured under ISO 13485:2016 certified quality management system