K599 Chemically Comp Agrobacterium Cells, Intact Genomics
Supplier: INTACT GENOMICS, INC MS
1087-06 1087-18
104090-454PK 375 USD
104090-454 104090-456
K599 Chemically Comp Agrobacterium Cells, Intact Genomics
Competent Cells
Intact Genomics (ig®) K599 Agrobacterium chemically competent cells are made from a specific strain of Agrobacterium rhizogenes, Agrobacterium rhizogenes (str R) pRi2659 (agropine type).
  • Useful for transgenic operations of corn, soybean (wild soybean), cotton, peanut, dandelion, cowpea and other plants
  • Transformation efficiency: ≥1×104  cfu/µg with control plasmid
  •  
Made from Agrobacterium rhizogenes (str R) pRi2659 (agropine type).
K599 was originally isolated from cucumber exhibiting hairy root disease symptoms and has been widely used for hairy root transformation.
Certifications:
Manufactured under ISO 13485:2016 certified quality management system.