ig® Thymidine Auxotrophic EHA105Dᵀʰʸ Agrobacterium Chemically Competent Cells, Intact Genomics
Supplier: INTACT GENOMICS, INC MS
1306-05 1306-15
104090-488PK 386.54 USD
104090-488 104090-490
ig® Thymidine Auxotrophic EHA105Dᵀʰʸ Agrobacterium Chemically Competent Cells, Intact Genomics
Competent Cells
Enables development of more efficient plant transformation systems.
  • Thymidine Auxotrophic as well as an atsD knockout mutant
  • Requires supplemental thymidine for growth
  • Reduced bacterial overgrowth during co-cultivation
  • Decreased need for antibiotics
Intact Genomics (ig®) Thymidine-Auxotrophic EHA105DThy Agrobacterium Chemically Competent Cells include modifications so that they will not grow unless 50 mg/L of thymidine is added to Minimal medium.
This prevents the bacteria from overgrowing plant tissues when used for plant transformation.