ig® Recombinase Polymerase Amplification (RPA) Kits v2
Supplier: INTACT GENOMICS, INC MS
3533 3534 3536
104090-544PK 398.08 USD
104090-544 104090-546 104090-548
ig® Recombinase Polymerase Amplification (RPA) Kits v2
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
The new improved IG® Recombinase Polymerase Amplification version 2 kit (IG® RPA v2) helps researchers who seek the advantages of isothermal DNA amplification to achieve faster, more discernible results.
  • IG® RPA v2 offers two key advantages over other kits:
  • More robust amplification
  • More specific amplification of the primary target, reduces background amplification of off-target sites
IG® Recombinase Polymerase Amplification (RPA) Kit v2 provides a mixture of the necessary enzymes to simplify and quicken the setup for your RPA reaction.
Certifications:
Manufactured under ISO 13485:2016 Quality Management certified standards.
Packaging:
Kit contains:
Enzyme mixture
2× RPA reaction master mix
280 mM Magnesium Acetate (MgOAc)
RPA positive control
Caution:
Store at −20 °C.