Glass Fiber Extraction Thimbles, Grade G400
Supplier: Sartorius
FT-1204-019090 FT-1204-022080 FT-1204-025100 FT-1204-030100 FT-1204-033094 FT-1204-043123 FT-1204-026060
77581-194PK 199.88 USD
77581-194 77581-196 77581-198 77581-202 77581-204 77581-206 77581-200
Glass Fiber Extraction Thimbles, Grade G400
Extractors Extraction Thimbles
These glass fiber thimble are primarily designed to fit the Soxhlet apparatus and potential applications include environmental monitoring (e.g. separation of dust, aerosols, gas or air streams) and food control (e.g. extraction of fats, emulsifiers or additives). 
  • Composed of glass fiber cellulose for uncontaminated results
  • Engineered for superior mechanical stability to withstand rigorous use
  • Glass fiber thimble offer remarkable resistance to high temperatures
  • Designed for a precise fit across a diverse range of extractors
  • Exhibits outstanding
More Product Information