Brady® i75 R6700 Ribbon for the i7500 Label Printer
Supplier: Brady Worldwide
176771 176772 176773
77775-060RL 0 USD
77775-060 77775-062 77775-064
Brady® i75 R6700 Ribbon for the i7500 Label Printer
Printer Ribbons
i75-Series ribbon with LabelSense™ Technology warns user if ribbon is incompatible with installed i75-Series label material. Automatically prints at pre-tested heat and speed, ensuring correct print quality on the first label with no setting adjustments or trial-and-error print testing.
  • Ribbon in cartridge format
  • Ribbin ink wind direction: ink side out
  • Polyester material
  • Halogen-free
i75-series ribbon is self-centering with ink-side always correctly facing the label. One-color ribbon for printing a single color onto a label. R6700 Series white wax/resin formulation offers excellent chemical, scratch, and smudge resistance when printed on the appropriate label material from Brady®.
Recommended for printing on dark-colored, heat-shrinkable tubing materials, such as black B-342 and B-344 PermaSleeve® wire markers from Brady®. Combine Brady label materials with their recommended ribbon series for superior legend performance.