NEBNext® Companion Module v2 for Oxford Nanopore Technologies® Ligation Sequencing
Supplier: New England Biolabs (NEB)
This module contains the NEBNext DNA repair, end repair and ligation reagents recommended for Oxford Nanopore Technologies® updated SQK-LSK114 singleplex ligation sequencing library prep protocols.
Includes Salt-T4® DNA Ligase & NEBNext FFPE DNA Repair Buffer v2
- Component volumes tailored for use with many SQK-LSK114 workflows
- Simplified ordering and inventory management
- Compatible with all devices: MinION®, GridION®, PromethION®, Flongle®
- No waste - no unnecessary buffers or excess reagents