Green Cellulose Sterilization Wraps, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
GHDSHEET4548 GHDSHEET1818 GHDSHEET6080 GHDSHEET0707 GHDSHEET4040 GHDSHEET1515 GHDSHEET1212 GHDSHEET4545 GHDSHEET0808 GHDSHEET5472 GHDSHEET3636 GHDSHEET01.5 GHDSHEET2424 GHDSHEET0404 GHDSHEET0606 GHDSHEET0909 GHDSHEET4848 GHDSHEET2020 GHDSHEET6060 GHDSHEET5454 GHDSHEET3030 GHDSHEET0303 GHDSHEET0202
77870-998CS 1189.88 USD
77870-998 77871-000 77871-002 77870-978 77870-980 77870-982 77870-958 77870-960 77870-962 77870-964 77870-966 77870-968 77870-970 77870-972 77870-974 77870-976 77870-984 77870-986 77870-988 77870-990 77870-992 77870-994 77870-996
Green Cellulose Sterilization Wraps, Keystone Cleanroom Products
Sterilization Wraps
When secured with autoclave tape or elasticized bands, these green GHD wrappers provide an effective microbial barrier from external sources of contamination.
  • Designed for steam sterilization; but also compatible with ETO or gamma irradiation
  • Treated to impart water and alcohol repellency
  • Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder
  • Sufficient microbial barrier
  • Superior bacterial barrier properties compared to linen
  • Sterilization tapes adhere firmly and are easy to remove
  • Ideal choice for light trays, small basins and towels
  • Resist tearing and punctures
  • Do not contain any natural latex rubber
  • Processed and packaged in ISO certified cleanrooms
Delivery information:
Certificates of Conformance sent with each shipment.
Packaging:
Packaged in sealed, labeled poly bags with lot number and date of manufacture.