Human IL-17A/F heterodimer cytokine ELISpot Kits
Supplier: Abcam
AB281965-1X96TESTS AB281965-5X96TESTS
ABCA_AB281965-1X96EA 620 USD
ABCA_AB281965-1X96 ABCA_AB281965-5X96
Human IL-17A/F heterodimer cytokine ELISpot Kits
Assays Immunological Assays
Human IL-17A/F heterodimer cytokine ELISpot Kit is a ELISpot Kit for the sensitive detection of Human IL-17A/F heterodimer cytokine production in a single cell suspension.