HydraGreen™ nucleic acid dyes
Supplier: ALADDIN SCIENTIFIC
HydraGreen™ Nucleic acid dyes
- High sensitivity: it is suitable for electrophoretic staining of various size fragments and has little effect on nucleic acid migration;
- High stability: suitable for preparing agarose gel by microwave or other heating methods;
- High signal-to-noise ratio: sample fluorescence signal is strong, background signal is low;
- Simple operation: no degradation in the process of prepreg and electrophoresis;
- Wide range of application: pre electrophoresis dyeing (gel dyeing) or post electrophoresis dyeing (bubble dyeing) can be selected.
HydraGreen ™, Water solution is nontoxic, non carcinogenic and has the same sensitivity as EB. It can be used to replace EB in nucleic acid electrophoresis experiment, dissolving in water instead of toxic DMSO.
Through the following tests, it is proved that there is no cancer risk
1. Ames test;
2. Mouse bone marrow micronucleus test;
3. Chromosome aberration test;
4. Penetration test of latex or nitrile gloves.
Packaging: Ambient