Anti-ZO1 tight junction protein Rabbit Monoclonal Antibody [clone: EPR19945-224]

Supplier: Abcam
AB221546-100UL AB221546-20UL AB221546-1ML
ABCA_AB221546-100UEA 796 USD
ABCA_AB221546-100U ABCA_AB221546-20UL ABCA_AB221546-1ML
Anti-ZO1 tight junction protein Rabbit Monoclonal Antibody [clone: EPR19945-224]
Antibodies
Rabbit monoclonal [EPR19945-224] to ZO1 tight junction protein.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: ZO1 tight junction protein
Clonality: Monoclonal
Clone: EPR19945-224
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR