Anti-NKCC1 Rabbit Monoclonal Antibody [clone: EPR26256-60]

Supplier: Abcam
AB303518-1ML AB303518-10UL AB303518-20UL AB303518-100UL
ABCA_AB303518-1MLEA 7319 USD
ABCA_AB303518-1ML ABCA_AB303518-10UL ABCA_AB303518-20UL ABCA_AB303518-100U
Anti-NKCC1 Rabbit Monoclonal Antibody [clone: EPR26256-60]
Antibodies
Rabbit monoclonal [EPR26256-60] to NKCC1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: NKCC1
Clonality: Monoclonal
Clone: EPR26256-60
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR