Anti-LRRK2 Rabbit Monoclonal Antibody [clone: UDD2 10(12)]

Supplier: Abcam
AB133450-100UL AB133450-1ML
ABCA_AB133450-100UEA 257 USD
ABCA_AB133450-100U ABCA_AB133450-1ML
Anti-LRRK2 Rabbit Monoclonal Antibody [clone: UDD2 10(12)]
Antibodies
Rabbit monoclonal [UDD2 10(12)] to LRRK2 (phospho S935).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: LRRK2
Clonality: Monoclonal
Clone: UDD2 10(12)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR