Anti-VGF Rabbit Monoclonal Antibody [clone: EPR27029-60]

Supplier: Abcam
AB313783-1ML AB313783-20UL AB313783-100UL
ABCA_AB313783-1MLEA 7144 USD
ABCA_AB313783-1ML ABCA_AB313783-20UL ABCA_AB313783-100U
Anti-VGF Rabbit Monoclonal Antibody [clone: EPR27029-60]
Antibodies
Rabbit monoclonal [EPR27029-60] to VGF.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: VGF
Clonality: Monoclonal
Clone: EPR27029-60
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR