Anti-SLC12A3 Rabbit Monoclonal Antibody [clone: EPR27106-48]

Supplier: Abcam
AB316191-20UL AB316191-100UL AB316191-1ML AB316191-10UL
ABCA_AB316191-20ULEA 276 USD
ABCA_AB316191-20UL ABCA_AB316191-100U ABCA_AB316191-1ML ABCA_AB316191-10UL
Anti-SLC12A3 Rabbit Monoclonal Antibody [clone: EPR27106-48]
Antibodies
Rabbit monoclonal [EPR27106-48] to SLC12A3.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SLC12A3
Clonality: Monoclonal
Clone: EPR27106-48
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR