Anti-Mast Cell Tryptase Rabbit Monoclonal Antibody [clone: EPR9522] (Biotin)

Supplier: Abcam
AB316948-10UL AB316948-100UL
ABCA_AB316948-10ULEA 232 USD
ABCA_AB316948-10UL ABCA_AB316948-100U
Anti-Mast Cell Tryptase Rabbit Monoclonal Antibody [clone: EPR9522] (Biotin)
Antibodies
Biotin Rabbit monoclonal [EPR9522] to Mast Cell Tryptase.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Mast Cell Tryptase
Clonality: Monoclonal
Clone: EPR9522
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR