Anti-ICOS Ligand/ICOSL Rabbit Monoclonal Antibody [clone: EPR20240]

Supplier: Abcam
AB209262-1ML AB209262-100UL
ABCA_AB209262-1MLEA 6273 USD
ABCA_AB209262-1ML ABCA_AB209262-100U
Anti-ICOS Ligand/ICOSL Rabbit Monoclonal Antibody [clone: EPR20240]
Antibodies
Rabbit monoclonal [EPR20240] to ICOS Ligand/ICOSL.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ICOS Ligand/ICOSL
Clonality: Monoclonal
Clone: EPR20240
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR