Anti-TRBC1 Rabbit Monoclonal Antibody [clone: SP305]

Supplier: Abcam
AB227699-100UL AB227699-1ML AB227699-500UL
ABCA_AB227699-100UEA 658 USD
ABCA_AB227699-100U ABCA_AB227699-1ML ABCA_AB227699-500U
Anti-TRBC1 Rabbit Monoclonal Antibody [clone: SP305]
Antibodies
Rabbit monoclonal [SP305] to TRBC1.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TRBC1
Clonality: Monoclonal
Clone: SP305
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR