Paragon Scientific Density 25 °C (Nominal: 1.1868 g/ml @ 25 °C)
Supplier: LGC Standards
ALK-DEN25-11
ALK-DEN25-11-500ML
77975-697EA
97
USD
77975-697
77975-698
Paragon Scientific Density 25 °C (Nominal: 1.1868 g/ml @ 25 °C)
Standards
Density Standards
- Shipping conditions: Room temperature
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...