Bard-Parker® Rib-Back™ Scalpel Blades
Supplier: StatLab
BP1322 BP1340
77907-832CS 359.91 USD
77907-832 77907-834
Bard-Parker® Rib-Back™ Scalpel Blades
Scalpels and Scalpel Assemblies Scalpel Blades
Bard-Parker® Rib-Back™ scalpel blades offer the superior cutting advantage of carbon steel technology.
  • Non-sterile blades
  • Individually wrapped
  • Packed 150 per box
Specially ground and carefully inspected, the exclusive Rib-Back™ blade design provides greater strength and control.