Anti-PPP3CA Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73633-100UL A73633-50UL
77848-289EA 330 USD
77848-289 77848-290
Anti-PPP3CA Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100

Type:
Antigen: PPP3CA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR