Anti-ARFGEF2 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A72314-100UG A72314-50UG
77846-023EA 330 USD
77846-023 77846-024
Anti-ARFGEF2 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: ARFGEF2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR