Anti-RAB35 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73101-100UL A73101-50UL
77847-398EA 330 USD
77847-398 77847-399
Anti-RAB35 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:10000, IHC: 1:50 to 1:200

Type:
Antigen: RAB35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR