Anti-AKT1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73600-100UL A73600-50UL
77848-234EA 330 USD
77848-234 77848-235
Anti-AKT1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:10000, WB: 1:500 to 1:1000

Type:
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR