Anti-MAST1 Rabbit Recombinant polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A72559-100UG A72559-50UG
77846-486EA 330 USD
77846-486 77846-487
Anti-MAST1 Rabbit Recombinant polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: MAST1
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR