Anti-NRN1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A67702-100UG A67702-50UG
77837-341EA 330 USD
77837-341 77837-342
Anti-NRN1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: NRN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR