Anti-SNX22 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A40355-100UG A40355-50UG
77807-487EA 330 USD
77807-487 77807-488
Anti-SNX22 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SNX22
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR