Anti-POMC Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A31642-50UG A31642-100UG
77794-814EA 225 USD
77794-814 77794-813
Anti-POMC Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: POMC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR